Lakeland CleanMax® Select Disposable Cleanroom Boot Covers, SMMS
Supplier: Lakeland
SBC903CMP-LGXL SBC903CMP-SMMD SBC903CSP-2X SBC903CSP-LGXL SBC903CSP-SMMD SBC903CMBP-2X SBC903CMBP-LGXL SBC903CMBP-SMMD SBC903CMP-2X
77188-502CS 346.45 CAD
77188-502 77188-504 77188-506 77188-508 77188-510 77188-494 77188-496 77188-498 77188-500
Lakeland CleanMax® Select Disposable Cleanroom Boot Covers, SMMS
Disposable Garments
The Lakeland CleanMax® Select boots covers are designed with breathable SMMS fabric that is ideal for ISO Class 5 to 8 cleanrooms.
  • Breathable SMMS fabric
  • Non-sterile and sterile options available
  • Tunnelized, latex-free elastic
  • Low linting
  • Elastic closure
  • 19" high
CleanMax® Select SMMS is available in sterile and non-sterile configurations. Clean-manufactured in Lakeland owned and operated certified cleanrooms, under strict process and environmental controls CleanMax® Select meets IEST particle category cleanliness levels. CleanMax® Select SMMS sterileMore Product Information