Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: AER311]

Supplier: ANTIBODIES.COM LLC
A316317-100
77179-548EA 1272.37 CAD
77179-548
Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: AER311]
Antibodies
Mouse monoclonal [AER311] antibody to Estrogen Receptor alpha for Flow Cytometry and IF with samples derived from Human, Bovine, Porcine, Mouse and Rat.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 0.5 to 1 µg/million cells, IF: 1 to 2 µg/ ml

Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: AER311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human;Bovine;Porcine;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR