Anti-Creatine Kinase MB Rabbit Recombinant Antibody [clone: CKBB/8608R]

Supplier: ANTIBODIES.COM LLC
A316206-100
77179-326EA 1272.37 CAD
77179-326
Anti-Creatine Kinase MB Rabbit Recombinant Antibody [clone: CKBB/8608R]
Antibodies
Recombinant rabbit monoclonal [CKBB/8608R] antibody to Creatine Kinase MB for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Creatine Kinase MB
Clonality: Recombinant
Clone: CKBB/8608R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR