Anti-Creatine Kinase MB Mouse Recombinant Antibody [clone: rCKBB/8841]

Supplier: ANTIBODIES.COM LLC
A316203-100
77179-320EA 1272.37 CAD
77179-320
Anti-Creatine Kinase MB Mouse Recombinant Antibody [clone: rCKBB/8841]
Antibodies
Recombinant mouse monoclonal [rCKBB/8841] antibody to Creatine Kinase MB for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Creatine Kinase MB
Clonality: Recombinant
Clone: rCKBB/8841
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR