Anti-Creatine Kinase MB Mouse Monoclonal Antibody [clone: CKBB/6567]

Supplier: ANTIBODIES.COM LLC
A316201-100
77179-316EA 1272.37 CAD
77179-316
Anti-Creatine Kinase MB Mouse Monoclonal Antibody [clone: CKBB/6567]
Antibodies
Mouse monoclonal [CKBB/6567] antibody to Creatine Kinase MB for ELISA, WB and IHC-P with samples derived from Human.

  • Validated Applications: ELISA, WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Creatine Kinase MB
Clonality: Monoclonal
Clone: CKBB/6567
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR