Anti-MASPIN Mouse Monoclonal Antibody [clone: SERPINB5/4978]

Supplier: ANTIBODIES.COM LLC
A315267-100
77177-448EA 1272.37 CAD
77177-448
Anti-MASPIN Mouse Monoclonal Antibody [clone: SERPINB5/4978]
Antibodies
Mouse monoclonal [SERPINB5/4978] antibody to MASPIN for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml

Type: Primary
Antigen: MASPIN
Clonality: Monoclonal
Clone: SERPINB5/4978
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR