Anti-Creatine Kinase MB Mouse Monoclonal Antibody [clone: CKBB/6567]

Supplier: ANTIBODIES.COM LLC
A314817-100
77176-548EA 1272.37 CAD
77176-548
Anti-Creatine Kinase MB Mouse Monoclonal Antibody [clone: CKBB/6567]
Antibodies
Mouse monoclonal [CKBB/6567] antibody to Creatine Kinase MB for ELISA, WB and IHC-P with samples derived from Human.

  • Validated Applications: ELISA, WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Creatine Kinase MB
Clonality: Monoclonal
Clone: CKBB/6567
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen synonyms creatine kinase muscle|Creatine Kinase MB|creatine kinase B|creatine kinase M|CKMM|brain creatine kinase|CKMB|creatine kinase brain|CKBB
Reactivity
Host
Gene ID UniprotID# P12277
Isotype
Immunogen Recombinant human full-length CKB protein
Form Liquid
Molecular weight 42 kDa
Purification Protein A/G chromatography
Concentration 200 µg/ml
Storage buffer Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
Storage temperature Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Shipping temperature Shipped on blue ice at 4 °C
Tested applications IHC-P

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR