LabFit™ Wall Cabinets
Supplier: TrippNT
53703 53704 53661
77135-384EA 3632.37 CAD
77135-384 77135-386 77135-302
LabFit™ Wall Cabinets
Furniture Casework
TrippNT® LabFit™ casework is constructed of half-inch polypropylene which is ideal for highly corrosive enviroments.
  • Non-metal construction
  • Resistant to chemicals and corrosion
  • Easy to clean and disinfect
  • Embedded magnetic door latches
  • Industry leading two year full replacement warranty
  • Made in USA
Polypropylene is a versatile plastic material that offers numberous beneftis for controlled environments. Its durability and resistnace to corrosion make it a practical choice. It can endure high temperatures and exposure to various chemicals, making it anMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable