OPTI-LYNX™ Holder Tube for sealing element
Supplier: Optimize
11-02327-AA
77100-358EA 157.02 CAD
77100-358
OPTI-LYNX™ Holder Tube for sealing element
Chromatography Supplies Chromatography Spare Parts
All OPTI-LYNX™ hardware can be purchased in kits or separately as needed for your application. OPTI-LYNX™ hardware is precision machined to maintain the highest quality and strictest tolerances.
  • For use with OPTI-LYNX™ hardware
  • Installs into holder cap to make connections with sealing elements
  • Modifies an OPTI-LYNX™ holder to accept sealing element inserts
Installs into holder cap to make connections with sealing elements.