OPTI-GUARD® Guard Columns, 1 mm
Supplier: Optimize
10-02-00010 10-02-00014 10-02-00018 10-02-00022 10-02-00026 10-02-00030 10-02-00034 10-02-01714 10-02-03532 10-02-00007 10-02-03211 10-02-03534 10-02-03536 10-02-03538 10-02-03680 10-02-03683 10-02-04708
77100-002PK 1249.04 CAD
77100-002 77100-004 77100-006 77100-008 77100-010 77100-012 77100-014 77100-028 89042-072 77100-000 89209-706 89209-708 89209-710 89209-712 89209-714 89209-716 10122-630
OPTI-GUARD® Guard Columns, 1 mm
Chromatography Columns
OPTI-GUARD® sets the standard for extremely low impact, easy to use pre-column protection. Designed for use with analytical (Int.Ø: 4.6, 3.0 mm) and narrow-bore (Int.Ø: 2.1, 1.0 mm) columns, the patented floating stem design automatically adjusts to any manufacturer's tube stop depth for a zero-dead-volume connection every time.
  • ZDV connection - automatically adjusts to any manufacturer's column
  • Requires no extra tubing for connection
  • Does not effect chromatography
  • No larger than a standard fitting
Connecting the OPTI-GUARD® requires no extra tubing,More Product Information