CryoFreeze® Cryogenic Vials
Supplier: SCIENTIFIC SPECIALTIES, INC.
6112-S0S 6121-S0S 6122-S0S 6131-S0S 6132-S0S 6141-S0S 6142-S0S 6212-S0S 6221-S0S 6222-S0S 6242-S0S
77093-426CS 117.99 CAD
77093-426 77093-428 77093-430 77093-432 77093-434 77093-436 77093-438 77093-440 77093-442 77093-444 77093-446
CryoFreeze® Cryogenic Vials
Vials
CryoFreeze® cryo vials offer a secure and convenient environment for your samples. Total quality assurance is central to the design and manufacturing philosophy. The CryoFreeze line brings a new dimension to ultra-low temperature storage.
  • Ideal for vapor-phase liquid nitrogen freezing
  • White writing area with black 100 µl graduations
  • Cap inserts are color coded
  • Round base or self standing Stargrip base
  • Sterile, certified free of DNas, RNase, DNA, PCR inhibitors and endotoxins
  • Internally threaded tube caps have a silicon-rubber O-Ring for a secure, leak-
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable