Anti-D6 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A99834-100
77076-604EA 765.04 CAD
77076-604
Anti-D6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CCBP2 for WB, IF and ELISA with samples derived from Human.

  • Validated Applications: WB, IF, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:1000

Type: Primary
Antigen: D6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR