Anti-Sucrose-Phosphate Synthase Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A101742-200
77074-970EA 974.39 CAD
77074-970
Anti-Sucrose-Phosphate Synthase Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Sucrose-Phosphate Synthase for WB and ELISA with samples derived from Plant.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: Sucrose-Phosphate Synthase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR