Anti-FH Goat Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A84362-100
77073-554EA 797.27 CAD
77073-554
Anti-FH Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to FH for ELISA, WB and IHC with samples derived from Human.

  • Validated Applications: ELISA, WB, IH

Type: Primary
Antigen: FH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR