Anti-Heat Labile Enterotoxin Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A101722-100
77061-392EA 893.81 CAD
77061-392
Anti-Heat Labile Enterotoxin Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Heat Labile Enterotoxin for WB and IP with samples derived from Heat Labile Enterotoxin and Cholera Toxin.

  • Validated Applications: WB, IP
  • Recommended Dilutions: WB: 1:2000 to 1:10000

Type: Primary
Antigen: Heat Labile Enterotoxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Heat Labile Enterotoxin;Cholera Toxin
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR