Anti-MMP16 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A95612-100
77050-996EA 765.04 CAD
77050-996
Anti-MMP16 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MMP-16 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000

Type: Primary
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol MMP16
Antigen name C8orf57
Environmentally Preferable
Antigen synonyms Matrix metalloproteinase 16|Membrane-type matrix metalloproteinase 3|MMP-16|Membrane type matrix metalloproteinase 3|Membrane-type-3 matrix metalloproteinase|DKFZp761D112|Membrane type 3 matrix metalloproteinase|Matrix metalloproteinase-16|MMP 16
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID UniprotID# P51512
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide derived from human MMP-16 (amino acids 551-600)
Form Liquid
Molecular weight 69 kDa
Purification Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
Storage buffer Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
Storage temperature Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Shipping temperature Shipped on blue ice at 4 °C
Tested applications IHC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR