Anti-14-3-3 beta + zeta Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A95556-100
77050-772EA 765.04 CAD
77050-772
Anti-14-3-3 beta + zeta Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to 14-3-3 beta for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000

Type: Primary
Antigen: 14-3-3 beta + zeta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol 14-3-3 beta + zeta
Environmentally Preferable
Antigen name 14 3 3 protein beta
Antigen synonyms HS1|KCIP1|YWHAZ|14 3 3 zeta|14 3 3 protein beta/alpha|YWHAB|14 3 3 protein zeta/delta|14-3-3 beta + zeta
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID UniprotID# P31946
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Synthetic peptide derived from human 14-3-3 beta (amino acids 41-90)
Form Liquid
Molecular weight 28 kDa
Purification Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
Storage buffer Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
Storage temperature Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Shipping temperature Shipped on blue ice at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR