Anti-Collagen VI Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A95139-100
77049-102EA 765.04 CAD
77049-102
Anti-Collagen VI Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Collagen VI alpha1 for IHC and ELISA with samples derived from Human and Mouse.

  • Validated Applications: IHC, ELISA
  • Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000

Type: Primary
Antigen: Collagen VI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol Collagen VI
Environmentally Preferable
Antigen name Alpha 1 (VI) chain (61 AA)
Antigen synonyms Collagen alpha 2(VI) chain|Collagen alpha 3(VI) chain|COL6A1|Collagen alpha-1(VI) chain|Collagen type VI alpha 1|COL6A2|CO6A1_HUMAN|Collagen type VI alpha 2|COL6A3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Gene ID UniprotID# P12109
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Immunogen Synthetic peptide derived from human Collagen VI alpha1 (amino acids 191-240)
Form Liquid
Molecular weight 108 kDa
Purification Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
Storage buffer Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
Storage temperature Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Shipping temperature Shipped on blue ice at 4 °C
Tested applications IHC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR