Anti-Histone H1oo Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A94705-100
77047-364EA 765.04 CAD
77047-364
Anti-Histone H1oo Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Histone H1 (acetyl Lys25) for WB, IHC and ELISA with samples derived from Human.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:1000

Type: Primary
Antigen: Histone H1oo
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR