Anti-ATP citrate lyase Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A93727-100
77043-450EA 765.04 CAD
77043-450
Anti-ATP citrate lyase Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATP-Citrate Lyase (phospho Ser454) for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000

Type: Primary
Antigen: ATP citrate lyase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR