Anti-EEF1A-pan (acetyl Lys41) Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A93542-100
77042-710EA 765.04 CAD
77042-710
Anti-EEF1A-pan (acetyl Lys41) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to EEF1A-pan (acetyl Lys41) for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:40000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR