Nexus™ Horizontal Clean Bench, 4' Wide
Supplier: Labconco
5340000 5340010 5340100 5340110 5340200 5340210 5340300 5340310 5340500 5341710 5341800 5341810 5340510 5340600 5340610 5340700 5340710 5340800 5340810 5341000 5341010 5341100 5341110 5341200 5341210 5341300 5341310 5341500 5341510 5341600 5341610 5341700
77037-800EA 16029.1 CAD
77037-800 77037-802 77037-804 77037-806 77037-808 77037-810 77037-812 77037-814 77037-816 77037-858 77037-860 77037-862 77037-818 77037-820 77037-822 77037-824 77037-826 77037-828 77037-830 77037-832 77037-834 77037-836 77037-838 77037-840 77037-842 77037-844 77037-846 77037-848 77037-850 77037-852 77037-854 77037-856
Nexus™ Horizontal Clean Bench, 4' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination. The Nexus provides an ISO 4 clean air environment.
More Product Information