Nexus™ Horizontal Clean Bench, 3' Wide
Supplier: Labconco
5331810 5331210 5331300 5331310 5331500 5331510 5331600 5331610 5331700 5331710 5331800 5330010 5330100 5330110 5330200 5330210 5330300 5330310 5330500 5330510 5330600 5330610 5330700 5330710 5330800 5330810 5331000 5331010 5331100 5331110 5331200
77037-798EA 18478.1 CAD
77037-798 77037-778 77037-780 77037-782 77037-784 77037-786 77037-788 77037-790 77037-792 77037-794 77037-796 77037-738 77037-740 77037-742 77037-744 77037-746 77037-748 77037-750 77037-752 77037-754 77037-756 77037-758 77037-760 77037-762 77037-764 77037-766 77037-768 77037-770 77037-772 77037-774 77037-776
Nexus™ Horizontal Clean Bench, 3' Wide
Fume Hoods and Enclosures Laminar Flow Cabinets
Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area.
  • Efficient unidirectional laminar airflow provided through CAP™ technology
  • Intelligent 5" touchscreen interface features intuitive monitoring capabilities
  • Reliable with long-life HEPA filters and maximum cleanability that stands up to disinfectants
The Nexus Horizontal clean bench is a laminar flow workstation designed to protect non-hazardous samples and processes from contamination. The Nexus provides an ISO 4 clean air environment.
More Product Information