Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE/iFluor® 750)

Supplier: AAT BIOQUEST INC
100411R0 100411R1 100411R2
77023-508EA 332.91 CAD
77023-508 77023-316 77024-984
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (PE/iFluor® 750)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: PE/iFluor® 750
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD4
Environmentally Preferable
Antigen name CD4 antigen
Antibody type Secondary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)/iFluor® 750
Clone RPA-T4
Reactivity Human
Host Mouse
Isotype IgG1
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); Minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR