Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Red)

Supplier: AAT BIOQUEST INC
103400V0 103400V1
77022-156EA 385.9 CAD
77022-156 77024-730
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Red)
Antibodies
4H11 is an anti-human monoclonal antibody that recognizes the CD34 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Red
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD34
Environmentally Preferable
Antigen name CD34 antigen
Antibody type Secondary
Clonality Monoclonal
Conjugation mFluor™ Red
Clone 4H11
Reactivity Human
Host Mouse
Isotype IgG1
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); Minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR