Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Green)

Supplier: AAT BIOQUEST INC
103400U0 103400U1
77024-072EA 385.9 CAD
77024-072 77021-108
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Green)
Antibodies
4H11 is an anti-human monoclonal antibody that recognizes the CD34 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Green
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR