Anti-CD19 Mouse Monoclonal Antibody (iFluor® 532) [clone: HIB19]

Supplier: AAT BIOQUEST INC
10192070 10192071
77023-554EA 385.9 CAD
77023-554 77022-130
Anti-CD19 Mouse Monoclonal Antibody (iFluor® 532) [clone: HIB19]
Antibodies
HIB19 is an anti-human monoclonal antibody that recognizes the CD19 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD19
Antigen name CD19 antigen
Environmentally Preferable
Antibody type Secondary
Clonality Monoclonal
Conjugation iFluor® 532
Clone HIB19
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR