Anti-CD28 Mouse Monoclonal Antibody (PE) [clone: CD28.2]

Supplier: AAT BIOQUEST INC
102801L0 102801L1 102801L2
77021-164EA 282.17 CAD
77021-164 77021-232 77022-800
Anti-CD28 Mouse Monoclonal Antibody (PE) [clone: CD28.2]
Antibodies
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD28
Antigen name CD28 antigen
Environmentally Preferable
Antibody type Secondary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone CD28.2
Reactivity Human
Host Mouse
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR