Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 680)

Supplier: AAT BIOQUEST INC
104410I0 104410I1
77022-586EA 385.9 CAD
77022-586 77021-044
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 680)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 680
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD44
Environmentally Preferable
Antigen name CD44 antigen
Antibody type Secondary
Clonality Monoclonal
Conjugation iFluor® 680
Clone IM7
Reactivity Human
Host Rat
Isotype
Purification Affinity purification
Storage temperature Refrigerated (2...8 °C); Minimize light exposure
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR