EZ DNA Methylation-Lightning Automation Kit
Supplier: Zymo Research
D5049
77020-292EA 480.6 CAD
77020-292
EZ DNA Methylation-Lightning Automation Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Automation-specific streamlined design for high-throughput bisulfite conversion of DNA for methylation analysis.
  • Ready to Use: Preassembled buffers allow for lightning-fast library preparation in as little as 4 hours without compromising quality
The EZ DNA Methylation-Lightning Automation Kit is a high-throughput DNA bisulfite conversion kit designed for streamlining automation workflows. It is recommended for several downstream Illumina® Infinium® products such as MethylationEPIC, HTS iSelect® Methyl Custom, and Mouse Methylation BeadChip Arrays. This automation-specific kit features optimized buffer volumes for consistent performance on all open platform liquid handlers and magnetic bead transfer automated systems.