Anti-Ustekinumab Mouse Monoclonal Antibody [clone: 9A5]
Supplier: Genscript
A02118-40
77007-640EA 243.16 CAD
77007-640
Anti-Ustekinumab Mouse Monoclonal Antibody [clone: 9A5]
Antibodies
Ustekinumab with trade name Stelara, is an FDA-approved drug to Crohn's disease, Ulcerative Colitis, Plaque Psoriasis and Psoriatic Arthritis, targeting both interleukins (IL)-12 and IL-23. Ustekinumab is a fully human immunoglobulin G1 kappa mAb that binds to IL-12 and IL-23. Ustekinumab binds to the p40 subunit common to IL-12 and IL-23 and prevents their interaction with the IL-12 receptor β1 subunit of the IL-12 and IL-23 receptor complexes, thereby disrupting the proinflammatory pathway. Anti-Ustekinumab Antibody (9A5), mAb, mouse is produced from a hybridoma resulting from theMore Product Information