Anti-Sarilumab Rabbit Polyclonal Antibody
Supplier: Genscript
A02122-40
77007-534EA 346.65 CAD
77007-534
Anti-Sarilumab Rabbit Polyclonal Antibody
Antibodies
Sarilumab with the trade name of Kevzara, is a human monoclonal IgG1 antibody. It binds to membrane bound and soluble interleukin 6 (IL-6) receptor forms. It was FDA-approved in for the treatment of Rheumatoid Arthritis (RA) in combination with methotrexate. GenScript anti-sarilumab antibody, pAb, rabbit is produced from sera of a rabbit immunized with Sarilumab.
  • High specificity
  • High affinity
  • High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research ofMore Product Information