Quick-DNA™ Plus Kits, Zymo Research
Supplier: Zymo Research
D4069 D4070 D4071 D4074 D4068 D4075 D4081 D4082 D4068T
77001-366EA 1261.9 CAD
77001-366 77001-368 77001-370 77001-372 76175-652 76197-090 76197-100 76197-126 76197-146
Quick-DNA™ Plus Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
DNA from any sample (cells, tissues, biological fluids and more)
  • Quick and easy: Simple 20 minute procedure
  • Highest yield: Recover 3× more DNA
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more
The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA ˃50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.
Order Now
SPECIFICATIONS
Description Quick-DNA™ 96 Plus Kit Quick-DNA™ Microprep Plus Kit Quick-DNA™ Miniprep Plus Kit Quick-DNA™ Magbead Plus Kit Quick-DNA™ Miniprep Plus Kit
Environmentally Preferable
Application
Target molecule DNA DNA DNA DNA DNA
Refine target molecule Total DNA Total DNA Total DNA Total DNA Total DNA
Sample type Blood,Biofluids (serum, plasma, saliva),Cultured cells,Human/animal cells and tissues Blood,Biofluids (serum, plasma, saliva),Cultured cells,Human/animal cells and tissues Blood,Biofluids (serum, plasma, saliva),Cultured cells,Human/animal cells and tissues Blood,Biofluids (serum, plasma, saliva),Cultured cells,Human/animal cells and tissues Blood,Biofluids (serum, plasma, saliva),Cultured cells,Human/animal cells and tissues
Method / Format Multiwell plate Column (silica-based) Column (silica-based) Magnetic beads Column (silica-based)
Prep size Mini Mini Mini Scalable Mini
Sample size ≤1×106 Cells or 5 mg Tissue ≤1×106 Cells or 5 mg Tissue ≤3×107 Cells or 125 mg Tissue ≤3×106 Cells or 25 mg Tissue ≤5×106 Cells or 25 mg Tissue
Sample capacity 5 µg 5 µg 125 µg 10 µg 25 µg
Yield The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled. The DNA binding capacity of the well is 5 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled. The DNA binding capacity of the column is 125 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled. Up to 10 µg per 50 µl of MagBinding Beads used The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.