Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ER506]

Supplier: ANTIBODIES.COM LLC
A251663-100
76967-378EA 877.69 CAD
76967-378
Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ER506]
Antibodies
Mouse monoclonal [ER506] antibody to Estrogen Receptor alpha for Flow Cytometry, WB and IHC-P with samples derived from Human.

  • Validated Applications: Flow Cytometry, WB, IHC-P
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: ER506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR