Anti-Glycophorin A Mouse Monoclonal Antibody [clone: JC159] (PE-Cyanine 5)

Supplier: ANTIBODIES.COM LLC
A122053-100
76965-752EA 716.69 CAD
76965-752
Anti-Glycophorin A Mouse Monoclonal Antibody [clone: JC159] (PE-Cyanine 5)
Antibodies
Mouse monoclonal [JC159] antibody to Glycophorin A (PE-Cyanine 5) for Flow Cytometry with samples derived from Rat and Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: Glycophorin A
Clonality: Monoclonal
Clone: JC159
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR