Anti-CD30 Mouse Monoclonal Antibody [Clone: Ber-H2 + CD30/412]

Supplier: ANTIBODIES.COM LLC
A254290-100 A254290-500
76963-026EA 603.88 CAD
76963-026 76954-980
Anti-CD30 Mouse Monoclonal Antibody [Clone: Ber-H2 + CD30/412]
Antibodies
Mouse monoclonal [Ber-H2 + CD30/412] antibody to CD30 for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1:100 to 1:200

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: Ber-H2 + CD30/412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human
Order Now

SPECIFICATIONS

Environmentally Preferable
Reactivity
Host
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR