Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ESR1/3564]

Supplier: ANTIBODIES.COM LLC
A251664-100
76959-726EA 877.69 CAD
76959-726
Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ESR1/3564]
Antibodies
Mouse monoclonal [ESR1/3564] antibody to Estrogen Receptor alpha for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: ESR1/3564
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR