Anti-CD80 Mouse Monoclonal Antibody [Clone: MEM-233] (PE (Phycoerythrin))

Supplier: ANTIBODIES.COM LLC
A85961-100
76958-578EA 716.69 CAD
76958-578
Anti-CD80 Mouse Monoclonal Antibody [Clone: MEM-233] (PE (Phycoerythrin))
Antibodies
Mouse monoclonal [MEM-233] antibody to CD80 (PE) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: MEM-233
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR