Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ER506] (PE (Phycoerythrin))

Supplier: ANTIBODIES.COM LLC
A251129-100
76957-892EA 877.69 CAD
76957-892
Anti-Estrogen Receptor alpha Mouse Monoclonal Antibody [clone: ER506] (PE (Phycoerythrin))
Antibodies
Mouse monoclonal [ER506] antibody to Estrogen Receptor alpha (PE) for Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 5 µl / million cells or 5 µl / 100 µl whole blood, IF: 1:50 to 1:100

Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Monoclonal
Clone: ER506
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR