Anti-Emerin Mouse Monoclonal Antibody [clone: EMD/2167]

Supplier: ANTIBODIES.COM LLC
A248425-100
76955-920EA 877.69 CAD
76955-920
Anti-Emerin Mouse Monoclonal Antibody [clone: EMD/2167]
Antibodies
Mouse monoclonal [EMD/2167] antibody to Emerin for WB, IF and IHC-P with samples derived from Human.

  • Validated Applications: WB, IF, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IF: 1 to 3 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Emerin
Clonality: Monoclonal
Clone: EMD/2167
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR