Anti-CD85j Mouse Monoclonal Antibody [Clone: GHI/75] (PE (Phycoerythrin))

Supplier: ANTIBODIES.COM LLC
A85829-100
76953-724EA 716.69 CAD
76953-724
Anti-CD85j Mouse Monoclonal Antibody [Clone: GHI/75] (PE (Phycoerythrin))
Antibodies
Mouse monoclonal [GHI/75] antibody to CD85j (PE) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD85j
Clonality: Monoclonal
Clone: GHI/75
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR