Anti-Bcl-6 Mouse Monoclonal Antibody [Clone: PCRP-BCL6-1E2]

Supplier: ANTIBODIES.COM LLC
A277776-100
76952-560EA 877.69 CAD
76952-560
Anti-Bcl-6 Mouse Monoclonal Antibody [Clone: PCRP-BCL6-1E2]
Antibodies
Mouse monoclonal [PCRP-BCL6-1E2] antibody to Bcl-6 for Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: Bcl-6
Clonality: Monoclonal
Clone: PCRP-BCL6-1E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR