Anti-EpCAM Mouse Monoclonal Antibody [clone: VU-1D9] (PE (Phycoerythrin))

Supplier: ANTIBODIES.COM LLC
A251139-100
76952-190EA 877.69 CAD
76952-190
Anti-EpCAM Mouse Monoclonal Antibody [clone: VU-1D9] (PE (Phycoerythrin))
Antibodies
Mouse monoclonal [VU-1D9] antibody to EpCAM (PE) for Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 5 µl / million cells or 5 µl / 100 µl whole blood, IF: 1:50 to 1:100

Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: VU-1D9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol EpCAM
Environmentally Preferable
Antigen name Cell surface glycoprotein Trop 2
Antigen synonyms CD326|Antigen identified by monoclonal|Cell surface glycoprotein Trop 1|AUA1|CD326 antigen|Adenocarcinoma-associated antigen|Adenocarcinoma associated antigen|323/A3|17 1A
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone VU-1D9
Reactivity Human
Host Mouse
Gene ID UniprotID# P16422
Isotype IgG1 kappa
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen Small cell lung carcinoma cells
Form Liquid
Molecular weight 40 - 43 kDa
Purification Protein A/G chromatography
Storage buffer Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
Storage temperature Store at 4...8 ℃. Antibody is stable for 24 months.
Shipping temperature Shipped on blue ice at 4 ℃

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR