Anti-Human IgE Mouse Monoclonal Antibody [Clone: BE5] (PE (Phycoerythrin))

Supplier: ANTIBODIES.COM LLC
A86634-100
76952-140EA 676.4 CAD
76952-140
Anti-Human IgE Mouse Monoclonal Antibody [Clone: BE5] (PE (Phycoerythrin))
Antibodies
Mouse monoclonal [BE5] antibody to Human IgE (PE) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: 2-5 µg/ml

Type: Primary
Antigen: Human IgE
Clonality: Monoclonal
Clone: BE5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol Human IgE
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone BE5
Reactivity Human
Host Mouse
Isotype IgG1
Flow cytometry Yes
Immunogen Purified human IgE
Form Liquid
Purification This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
Concentration 0.1 mg/ml
Storage buffer Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
Storage temperature Store in the dark at 2...8 °C. Do not freeze!
Shipping temperature Shipped on blue ice at 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR