LISA Safety Waste Cap
Supplier: SCAT AMERICAS, INC.
350090 350095 350099 350100 350110 350120 350121 450045 450060 450100 450110 450120 350045 350050 350051 350053 350055 350060 350063 350065 350070 350083
76948-946EA 425.9 CAD
76948-946 76948-948 76948-950 76948-952 76948-954 76948-956 76948-958 76948-974 76948-976 76948-978 76948-980 76948-982 76948-926 76948-928 76948-930 76948-932 76948-934 76948-936 76948-938 76948-940 76948-942 76948-944
LISA Safety Waste Cap
Closures Safety Caps and Safety Waste Caps
The first modular system for HPLC waste. Workplace safety and environmental protection with the proven technology, providing maximum flexibility. Simply add connections and components individually as required. The modular system grows with the user, allowing solvents, acids and alkalis to be collected safely.
  • Satellite blind plug extends and attaches various components, such as a funnel
  • Tube connector allows connection of larger tubes with 5 to 11.5 mm Int.Ø
  • Satellite made of PTFE for optimal chemical resistance
  • 360° freely rotatable screw cap; easy
More Product Information