Microscope Slides, Non-Charged, Inkjet, 45° Corners
Supplier: Mopec
SL445-PK-BX SL445-PK-CS SL445-TN-BX SL445-TN-CS SL445-WH-BX SL445-WH-CS SL445-YL-BX SL445-YL-CS SL445-BL-BX SL445-BL-CS SL445-GR-BX SL445-GR-CS SL445-LI-BX SL445-LI-CS SL445-PE-BX SL445-PE-CS
76948-478BX 58.13 CAD
76948-478 76948-480 76948-482 76948-484 76948-486 76948-488 76948-490 76948-492 76948-462 76948-464 76948-466 76948-468 76948-470 76948-472 76948-474 76948-476
Microscope Slides, Non-Charged, Inkjet, 45° Corners
Slides Microscope Slides
High-quality, uncharged microscope slides with color coding.
  • For Inkjet
  • 1×3" slides, 1 mm thickness
  • 100% guaranteed against sticking and contamination
  • 45 degree corners
  • Uniform flat paint for perfect barcodes
Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac. The packaging options include 1 box of 72 slides and 1 case of 1440 slides.