Rapid Response™ Fentanyl Test Strips (Liquid/Powder)
Supplier: BTNX INC.
FYL-18S7-100 FYL-18S7-100
76945-146EA 157.78 CAD
76945-146 CA76945-146
Rapid Response™ Fentanyl Test Strips (Liquid/Powder)
Drug Tests
The Rapid Response™ Fentanyl test strips (Liquid/Powder) detects fentanyl in suspicious substances on surfaces and liquids from suspicious receptables at a cut-off concentration of 200 ng/ml.
  • Sample: Liquid/Powder
  • Detects fentanyl at a cut-off concentration of 200 ng/ml
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips
  • Sensitive to fentanyl and analogues: Carfentanil, Butyryl Fentanyl, p-Fluoro Fentanyl, Acetyl Fentanyl, Fentanyl, Furanyl Fentanyl, Valeryl Fentanyl,
More Product Information