QRP® Qualakote® Nylon/Carbon Fiber ESD Gloves with Polyurethane Fingertip Coating
Supplier: PIP
TDESDNYM TDESDNYS TDESDNYXL TDESDNYXS TDESDNY-2X TDESDNYL
76945-138CS 532.33 CAD
76945-138 76945-136 76945-142 76945-134 76945-144 76945-140
QRP® Qualakote® Nylon/Carbon Fiber ESD Gloves with Polyurethane Fingertip Coating
Gloves
Seamless knit nylon/carbon fiber electrostatic dissipative (ESD) gloves with polyurethane coated grip on fingertips.
  • Seamless knit gloves provide great dexterity
  • Polyurethane coated fingertips provide a secure grip in dry and slightly wet/oily conditions
  • High dexterity and tactile sensitivity
  • Reusable and launderable
Ideally suited for use in circuit board manufacturing, munitions, electronics assembly, and aerospace applications. The electrostatic dissipative (ESD) fiber yarns blended with low-lint nylon reduce static build-up on the glove surfaceMore Product Information