CleanTeam® Nylon/Carbon Fiber ESD Gloves
Supplier: PIP
40-6410/XL 40-6410/XXL 40-6411/L 40-6411/S
76944-952DZ 44.21 CAD
76944-952 76944-954 76944-958 76944-956
CleanTeam® Nylon/Carbon Fiber ESD Gloves
Gloves
Seamless knit nylon/carbon fiber electrostatic dissipative (ESD) gloves, available with or without raised PVC dot grip enhancement.
  • Electrostatic dissipative (ESD) carbon fiber yarns blended with low-lint nylon
  • Reduced static build-up on the glove surface for improved performance in electronics assembly
  • Seamless knit gloves provide great dexterity
  • Reusable and launderable
Ideally suited for use in circuit board manufacturing, munitions, electronics assembly, and aerospace applications. Gloves featuring raised PVC dots provide enhanced grip and extraMore Product Information