PIP® AR/FR Dual-Certified Coverall with Zipper
Supplier: PIP
385-FRSC-NV/3X 385-FRSC-NV/4X 385-FRSC-NV/5X 385-FRSC-NV/L 385-FRSC-NV/M 385-FRSC-NV/S 385-FRSC-NV/XL 385-FRSC-KH/2X 385-FRSC-KH/3X 385-FRSC-KH/4X 385-FRSC-KH/5X 385-FRSC-KH/L 385-FRSC-KH/M 385-FRSC-KH/S 385-FRSC-KH/XL 385-FRSC-NV/2X
76944-926CS 2091.95 CAD
76944-926 76944-928 76944-930 76944-920 76944-918 76944-916 76944-922 76944-908 76944-910 76944-912 76944-914 76944-904 76944-902 76944-900 76944-906 76944-924
PIP® AR/FR Dual-Certified Coverall with Zipper
Workwear
Coveralls are manufactured from a cotton/nylon blend that provides comfort and durability throughout extended wear.
  • Arc rating: ATPV 9.2 cal/cm²
  • PPE Category: Two
  • 90% cotton/10% nylon construction
  • Two-way front brass zipper on aramid tape
  • Storm guard over front zipper
  • Protective shirt collar; side elasticized waist
  • Arm cuffs with two position snap closure
All coveralls feature two chest flap pockets with snap closure, two waist and two rear hip patch pockets, a pen pocket on the left sleeve, and a tool pocket on the leg. All metal components areMore Product Information